Learn More
GENERAL BIOPHYSICS LLC Krypton Gas Analyzer Model B
Catalog No. NC3361687
Encompass
Thermoconductivity Gas Analyzer for Krypton (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.
Catalog No. NC3361687 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAKRO2B
Thermoconductivity Gas Analyzer for Krypton (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.