Learn More
Research Products International Corp Krystal Assay Plates, 96x350ul, Black, 100/pk
SDP
Catalog No. 501976994
Encompass
Krystal assay plates feature clear well bottoms for use with instruments that read above or below the plates. Standard 96 well plates have a well volume of 300ul. Tissue culture treated and individually wrapped with lid.
Catalog No. 50-197-6994 Supplier Research Products International Corp Supplier No. 141361
Krystal assay plates feature clear well bottoms for use with instruments that read above or below the plates. Standard 96 well plates have a well volume of 300ul. Tissue culture treated and individually wrapped with lid.