Learn More
Chemglass Life Sciences Krystal Microplate, 384 Well, White, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair
SDP
Catalog No. 501944003
Encompass_Preferred
Krystal Microplate, 384 Well, White, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair. The Krystal 384-well microplate range incorporates novel square wells with a large liquid capacity of 120µl per well, increasing the achievable sensitivity of most HTS assays. Using a proprietary manufacturing technique, the Krystal 384 offers market leading plate flatness (+/- 0.1mm tolerance) that translates into a significant increase in measurement precision and elimination of read errors when performing cell-based assays using fluorescent or luminescent imaging. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.
Catalog No. 50-194-4003 Supplier Chemglass Life Sciences Supplier No. CLS3579120WT
Krystal Microplate, 384 Well, White, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair. The Krystal 384-well microplate range incorporates novel square wells with a large liquid capacity of 120µl per well, increasing the achievable sensitivity of most HTS assays. Using a proprietary manufacturing technique, the Krystal 384 offers market leading plate flatness (+/- 0.1mm tolerance) that translates into a significant increase in measurement precision and elimination of read errors when performing cell-based assays using fluorescent or luminescent imaging. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.