Learn More
Chemglass Life Sciences Krystal Microplate, 384 Well, Black, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair
SDP
Catalog No. 501944004
Encompass_Preferred
Krystal Microplate, 384 Well, Black, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair. The Krystal 384-well microplate range incorporates novel square wells with a large liquid capacity of 120µl per well, increasing the achievable sensitivity of most HTS assays. Using a proprietary manufacturing technique, the Krystal 384 offers market leading plate flatness (+/- 0.1mm tolerance) that translates into a significant increase in measurement precision and elimination of read errors when performing cell-based assays using fluorescent or luminescent imaging. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.
Catalog No. 50-194-4004 Supplier Chemglass Life Sciences Supplier No. CLS3579120BT
Add to Cart
Add to Cart
Krystal Microplate, 384 Well, Black, Square Wells, Tissue Culture Treated, Individually Wrapped, Sterile, Porvair. The Krystal 384-well microplate range incorporates novel square wells with a large liquid capacity of 120µl per well, increasing the achievable sensitivity of most HTS assays. Using a proprietary manufacturing technique, the Krystal 384 offers market leading plate flatness (+/- 0.1mm tolerance) that translates into a significant increase in measurement precision and elimination of read errors when performing cell-based assays using fluorescent or luminescent imaging. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.