Learn More
Chemglass Life Sciences Krystal Microplate, 96 Well, Black, Non-Sterile, Assay, Porvair
SDP
Catalog No. 5000980736
Encompass_Preferred
Opaque walls to prevent well-to-well crosstalk. Clear plate bottom permits direct microscopic viewing. Thickness of bottom 0.75mm. For use with top or bottom reading instruments. Standard ANSI/SLAS format. Well volume of 350µl. Constructed from ultra-pure grade polystyrene. Clear bottom 96-well Krystal plates are optimized for fluorescence and luminescence. Proprietary ultra-sonic welding is used to assemble the clear bases to the solid top plate in our clean room. This design provides flat, optically clear bases to each well with no leakage, enabling excellent accuracy and reproducibility of photometric readings. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.
Catalog No. 50-009-80736 Supplier Chemglass Life Sciences Supplier No. CLS3577350B
Opaque walls to prevent well-to-well crosstalk. Clear plate bottom permits direct microscopic viewing. Thickness of bottom 0.75mm. For use with top or bottom reading instruments. Standard ANSI/SLAS format. Well volume of 350µl. Constructed from ultra-pure grade polystyrene. Clear bottom 96-well Krystal plates are optimized for fluorescence and luminescence. Proprietary ultra-sonic welding is used to assemble the clear bases to the solid top plate in our clean room. This design provides flat, optically clear bases to each well with no leakage, enabling excellent accuracy and reproducibility of photometric readings. All tissue-culture treated plates are supplied lidded and sterile, in individual bags.