Learn More
TMC Industries KRYTOX EG 2000 Grease, 2 ounce tube
SDP
Catalog No. NC3661216
Encompass
Krytox EG 2000 are similar to aerospace-quality fluorinated grease equivalent to 240AC but without the MIL-SPEC certification, making it significantly more economical. Intended for deep vacuum applications. It is a white buttery grease that uses Krytox 2000 PTFE thickener that has a melting point of 617ºF.The base is Krytox 143 series oil a clear odorless perfluorinated polyether ( PFPE ) synthetic that is nonflammable, nontoxic and nonreactive safe in oxygen and chemical environments. Because of its exceptionally low vapor pressure, in most terrestrial applications this grease will literally never dry out.
Catalog No. NC3661216 Supplier TMC Industries Supplier No. 36194
Krytox EG 2000 are similar to aerospace-quality fluorinated grease equivalent to 240AC but without the MIL-SPEC certification, making it significantly more economical. Intended for deep vacuum applications. It is a white buttery grease that uses Krytox 2000 PTFE thickener that has a melting point of 617ºF.The base is Krytox 143 series oil a clear odorless perfluorinated polyether ( PFPE ) synthetic that is nonflammable, nontoxic and nonreactive safe in oxygen and chemical environments. Because of its exceptionally low vapor pressure, in most terrestrial applications this grease will literally never dry out.