Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC Krytox GPL 227 | 2oz
SDP
Catalog No. NC1840671
Encompass_Preferred
Krytox GPL 227 grease provides protection from rusting, high temperature corrosion, and antiwear. It has excellent lubrication over a broad temperature range. Krytox GPL 227 is nonflammable, oxygen compatible, and chemically inert. Krytox allows extended lubrication intervals and longer equipment life.
Catalog No. NC1840671 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. GPL2272OZ
Krytox GPL 227 grease provides protection from rusting, high temperature corrosion, and antiwear. It has excellent lubrication over a broad temperature range. Krytox GPL 227 is nonflammable, oxygen compatible, and chemically inert. Krytox allows extended lubrication intervals and longer equipment life.