Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC Krytox™ LVP | 8oz
SDP
Catalog No. NC1393012
Encompass_Preferred
Krytox LVP high-vacuum grease performs over extremes of temperature and pressure that hydrocarbon, silicone, and chlorofluorocarbon greases can’t match. From –15–300°C (0–570°F), at vapor pressures as low as 10–13 mmHg, and in even the most chemically severe environments, Krytox LVP grease gives dependable, proven performance.
Catalog No. NC1393012 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. LVP8OZ
Krytox LVP high-vacuum grease performs over extremes of temperature and pressure that hydrocarbon, silicone, and chlorofluorocarbon greases can’t match. From –15–300°C (0–570°F), at vapor pressures as low as 10–13 mmHg, and in even the most chemically severe environments, Krytox LVP grease gives dependable, proven performance.