Learn More
Sigma Aldrich Fine Chemicals Biosciences KrytoxR 240AC lubricant pk1EA
Catalog No. 502904729
Encompass
fluorinated grease. Multi-purpose lubricant chemically inert non-flammable.Useful temp range -30 to 550 fInsoluble in most common solvents
Catalog No. 50-290-4729 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1230721EA
fluorinated grease. Multi-purpose lubricant chemically inert non-flammable.Useful temp range -30 to 550 fInsoluble in most common solvents