Learn More
Sigma Aldrich Fine Chemicals Biosciences KrytoxR LVP grease weight1EA
Catalog No. 502901343
Encompass
Similiar to Krytox 240AAC except used as high-vacuum grease. 2Aoz. tube. R 36/37/38 S 26-36
Catalog No. 50-290-1343 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2735461EA
Similiar to Krytox 240AAC except used as high-vacuum grease. 2Aoz. tube. R 36/37/38 S 26-36