Learn More
Sigma Aldrich Fine Chemicals Biosciences KrytoxR vacuum oil grade 1500G
Catalog No. 502901008
Encompass
DuPont Krytox vacuum pump fluids are used in applications where conventional vacuum pump oils cause safety waste disposal and maintenance problems. They are nonflammable and eliminate the chance of fire in pumps. They are nonreactive and safe to use in oxygen systems. This gives Krytox superior stability as a vacuum pump fluid. Krytox vacuum fluids are precisely distilled to provide low vapor pressures and give superior performance.NonflammableNonreactivefree from acetal groupsRecyclable
Catalog No. 50-290-1008 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z678465500G
DuPont Krytox vacuum pump fluids are used in applications where conventional vacuum pump oils cause safety waste disposal and maintenance problems. They are nonflammable and eliminate the chance of fire in pumps. They are nonreactive and safe to use in oxygen systems. This gives Krytox superior stability as a vacuum pump fluid. Krytox vacuum fluids are precisely distilled to provide low vapor pressures and give superior performance.NonflammableNonreactivefree from acetal groupsRecyclable