Learn More
Enzo Life Sciences KS-176 (2mg)
SDP
Supplier:  Enzo Life Sciences ALX270512M002
Encompass_Preferred
A potent specific inhibitor of Breast Cancer Resistance Protein (BCRP) / ATP-binding cassette sub-family G member 2 (ABCG2). IC50=1.39µM Alternative name: N-(4-(2-hydroxyethyl)phenyl)-2-(4-nitrobenzamido)benzamide. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8631