Learn More
Hach Company KS100 KCl Solution, Saturated, 500 mL (Radiometer Analytical)
Catalog No. 501997549 Shop All Hach Company Products
Encompass
Filling and storage solution for combined/reference electrodes.
Catalog No. 50-199-7549 Supplier Hach Company Supplier No. C20C300
Filling and storage solution for combined/reference electrodes.