Learn More
Hach Company KS110 KCl Solution 3M 500 mL
Catalog No. 5000207019 Shop All Hach Company Products
Encompass
KS110 KCl Solution-3M-500 mL (Radiometer Analytical)Filling and storage solution for combined/reference electrodes
Catalog No. 50-002-07019 Supplier Hach Company Supplier No. C20C320
KS110 KCl Solution-3M-500 mL (Radiometer Analytical)Filling and storage solution for combined/reference electrodes