Learn More
Hach Company KS120 KCl Sol Sat. Sat. w/AgCl
Catalog No. 5000207045 Shop All Hach Company Products
Encompass
KS120 KCl Solution Sat -Saturated with AgCl-500 mL (Radiometer Analytical)Filling and storage solution for combined/reference electrodes
Catalog No. 50-002-07045 Supplier Hach Company Supplier No. C20C310
KS120 KCl Solution Sat -Saturated with AgCl-500 mL (Radiometer Analytical)Filling and storage solution for combined/reference electrodes