Learn More
TARGETMOL CHEMICALS INC KS370G 5MG
SDP
Catalog No. 502580817
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. KS370G (Caffeic Acid Phenethyl Amide) inhibits UUO-induced renal fibrosis marker expression. KS370G is an orally active hypoglycemic and cardioprotective agent that reduces collagen deposition in obstructed kidneys and significantly decreases renal expression of inflammatory chemokines/adhesion molecules and monocyte markers and improves left ventricular hypertrophy and function in the heart of pressure-overloaded mice.KS370G can be used to study renal obstructive nephropathy. purity: 99%
Catalog No. 50-258-0817 Supplier TARGETMOL CHEMICALS INC Supplier No. T242705MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. KS370G (Caffeic Acid Phenethyl Amide) inhibits UUO-induced renal fibrosis marker expression. KS370G is an orally active hypoglycemic and cardioprotective agent that reduces collagen deposition in obstructed kidneys and significantly decreases renal expression of inflammatory chemokines/adhesion molecules and monocyte markers and improves left ventricular hypertrophy and function in the heart of pressure-overloaded mice.KS370G can be used to study renal obstructive nephropathy. purity: 99%