Learn More
Medchemexpress LLC Ksc-34 | 2226201-97-2 | 99.9% | 429.95 | C21H28ClN7O | 1 ML
SDP
Catalog No. 5000226767
Encompass_Preferred
KSC-34 is a covalent modifier and selective inhibitor of protein disulfide isomerase A1 (PDIA1) intended for research use. It is available as a ready-to-use DMSO solution and in multiple solid pack sizes, and is supplied with detailed solubility and storage recommendations.
  • Selective inhibition of PDIA1 a-site with IC50 3.5 μM.
  • Covalent modifier suitable for biochemical and cellular studies.
  • High purity (99.94%) to support reproducible results.
  • Supplied as a 10 mM solution in DMSO (1 mL) and in several solid pack sizes.
  • High DMSO solubility (100 mg/mL); ultrasonic assistance recommended.
  • Defined storage recommendations for solid and solution to preserve stability.
Catalog No. 50-002-26767 Supplier Medchemexpress LLC Supplier No. HY11757010MM 1ML
KSC-34 is a covalent modifier and selective inhibitor of protein disulfide isomerase A1 (PDIA1) intended for research use. It is available as a ready-to-use DMSO solution and in multiple solid pack sizes, and is supplied with detailed solubility and storage recommendations.
  • Selective inhibition of PDIA1 a-site with IC50 3.5 μM.
  • Covalent modifier suitable for biochemical and cellular studies.
  • High purity (99.94%) to support reproducible results.
  • Supplied as a 10 mM solution in DMSO (1 mL) and in several solid pack sizes.
  • High DMSO solubility (100 mg/mL); ultrasonic assistance recommended.
  • Defined storage recommendations for solid and solution to preserve stability.