Learn More
Medchemexpress LLC KSC-34 | 2226201-97-2 | 99.9% | 429.95 | C21H28ClN7O | 10 MG
SDP
Catalog No. 5000227850
Encompass_Preferred
KSC-34 is a covalent modifier and selective a-site inhibitor of protein disulfide isomerase A1 (PDIA1), provided as a high-purity solid for biochemical and cell-based research. It contains an alkyne group enabling click chemistry and can be formulated for in vitro or in vivo use.
  • Covalent modifier of PDIA1 with a-site selectivity.
  • IC50 of 3.5 μM at the PDIA1 a active site.
  • Alkyne handle compatible with copper-catalyzed azide-alkyne cycloaddition.
  • Reported high purity (about 99.94%).
  • Soluble in DMSO; manufacturer provides in vitro and in vivo formulation guidance.
Catalog No. 50-002-27850 Supplier Medchemexpress LLC Supplier No. HY11757010MG
KSC-34 is a covalent modifier and selective a-site inhibitor of protein disulfide isomerase A1 (PDIA1), provided as a high-purity solid for biochemical and cell-based research. It contains an alkyne group enabling click chemistry and can be formulated for in vitro or in vivo use.
  • Covalent modifier of PDIA1 with a-site selectivity.
  • IC50 of 3.5 μM at the PDIA1 a active site.
  • Alkyne handle compatible with copper-catalyzed azide-alkyne cycloaddition.
  • Reported high purity (about 99.94%).
  • Soluble in DMSO; manufacturer provides in vitro and in vivo formulation guidance.