Learn More
Medchemexpress LLC Ksc-34 | 2226201-97-2 | 99.9% | 429.95 | C21H28ClN7O | 25 MG
SDP
Catalog No. 5000226952
Encompass_Preferred
KSC-34 is a covalent modifier and selective a-site inhibitor of protein disulfide isomerase A1 (PDIA1), supplied as a high-purity research reagent. It contains an alkyne functional group enabling copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is provided in multiple solid and solution formats for laboratory use.
  • Selective PDIA1 a-site inhibitor (IC50 ≈ 3.5 μM).
  • Contains an alkyne group for click chemistry applications.
  • High purity suitable for research use (~99.9%).
  • Available as solids (including 25 mg) and as a DMSO solution.
  • Supplied with supporting datasheet and SDS documentation.
Catalog No. 50-002-26952 Supplier Medchemexpress LLC Supplier No. HY11757025MG
KSC-34 is a covalent modifier and selective a-site inhibitor of protein disulfide isomerase A1 (PDIA1), supplied as a high-purity research reagent. It contains an alkyne functional group enabling copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is provided in multiple solid and solution formats for laboratory use.
  • Selective PDIA1 a-site inhibitor (IC50 ≈ 3.5 μM).
  • Contains an alkyne group for click chemistry applications.
  • High purity suitable for research use (~99.9%).
  • Available as solids (including 25 mg) and as a DMSO solution.
  • Supplied with supporting datasheet and SDS documentation.