Learn More
Selleck Chemical LLC Ksg hydrochloride hydrate
SDP
Catalog No. 5000003406
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate an antibiotic which binds both the 30S and 70S ribosome specifically inhibits translation initiation of canonical but not of leaderless messenger RNAs
Catalog No. 50-000-03406 Supplier Selleck Chemical LLC Supplier No. S596825MG
Kasugamycin (Ksg) hydrochloride hydrate an antibiotic which binds both the 30S and 70S ribosome specifically inhibits translation initiation of canonical but not of leaderless messenger RNAs