Learn More
Cayman Chemical KSL-128114 (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502915340
Encompass_Preferred
A peptide syntenin inhibitor (Ki = 300 nM); reduces the percent viability of A2058 melanoma and patient-derived GBM cells in a concentration-dependent manner; increases animal survival time in a PDX mouse model of GBM using patient-derived GBM cells co-cultured with KSL-128114 prior to intracranial implantation at 50 µM
Catalog No. 50-291-5340 Supplier Cayman Chemical Supplier No. 3698410 MG
A peptide syntenin inhibitor (Ki = 300 nM); reduces the percent viability of A2058 melanoma and patient-derived GBM cells in a concentration-dependent manner; increases animal survival time in a PDX mouse model of GBM using patient-derived GBM cells co-cultured with KSL-128114 prior to intracranial implantation at 50 µM