Learn More
Sigma Aldrich Fine Chemicals Biosciences EmbryoMax Advanced KSOM Embryo Medium | 50mL
Catalog No. NC1807904
Encompass
EmbryoMax Advanced KSOM Medium can be used as a single medium to harvest and culture preimplantation mouse embryos, replacing the normal 2-step system where one medium is used for embryo harvest and another is used for culture.
Catalog No. NC1807904 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MR101D
EmbryoMax Advanced KSOM Medium can be used as a single medium to harvest and culture preimplantation mouse embryos, replacing the normal 2-step system where one medium is used for embryo harvest and another is used for culture.