Learn More
TRIBIOSCIENCE INC KSOM Mouse Embryo Medium With AA, 50 ML
SDP
Catalog No. NC3454604
Encompass_Preferred
Tribioscience’s KSOM Mouse Embryo Medium With AA is a potassium simplex optimized medium for in vitro culture of mouse oocytes. The Size of medium is 50 mL/ bottle.
Catalog No. NC3454604 Supplier TRIBIOSCIENCE INC Supplier No. TBS8071AA
Tribioscience’s KSOM Mouse Embryo Medium With AA is a potassium simplex optimized medium for in vitro culture of mouse oocytes. The Size of medium is 50 mL/ bottle.