Learn More
Medchemexpress LLC KSPWFTTL TFA | 99.3% | 1093.15 | 5 MG
SDP
Catalog No. 5000399284
Encompass_Preferred
KSPWFTTL TFA is an immunodominant Kb-restricted epitope derived from the p15E transmembrane protein. This peptide can restore susceptibility of a tumor line to anti-AKR/Gross MuLV cytotoxic T lymphocytes. It is derived from the retroviral p15 TM envelope protein.
  • Immunodominant Kb-restricted epitope.
  • Derived from p15E transmembrane protein.
  • Restores susceptibility of tumor line to anti-AKR/Gross MuLV cytotoxic T lymphocytes.
  • Makes cytolytic T-lymphocyte (CTL)-insusceptible clonal line susceptible to lysis by antiviral CTL when pulsed with the peptide.
Catalog No. 50-003-99284 Supplier Medchemexpress LLC Supplier No. HYP3333A5MG
KSPWFTTL TFA is an immunodominant Kb-restricted epitope derived from the p15E transmembrane protein. This peptide can restore susceptibility of a tumor line to anti-AKR/Gross MuLV cytotoxic T lymphocytes. It is derived from the retroviral p15 TM envelope protein.
  • Immunodominant Kb-restricted epitope.
  • Derived from p15E transmembrane protein.
  • Restores susceptibility of tumor line to anti-AKR/Gross MuLV cytotoxic T lymphocytes.
  • Makes cytolytic T-lymphocyte (CTL)-insusceptible clonal line susceptible to lysis by antiviral CTL when pulsed with the peptide.