Learn More
Selleck Chemical LLC KT 474-E1655-1MG
SDP
Catalog No. 503134500
Encompass_Preferred
KT-474 (SAR444656) is a selective small-molecule degrader of IRAK4 in development for the treatment of TLR/IL-1R driven autoimmune diseases KT-474 potently downregulates R848 (TLR7/8) and lipopolysaccharide (LPS)-induced IL-6 and IL-8 production in peripheral blood mononuclear cells (PBMCs)
Catalog No. 50-313-4500 Supplier Selleck Chemical LLC Supplier No. E16551MG
KT-474 (SAR444656) is a selective small-molecule degrader of IRAK4 in development for the treatment of TLR/IL-1R driven autoimmune diseases KT-474 potently downregulates R848 (TLR7/8) and lipopolysaccharide (LPS)-induced IL-6 and IL-8 production in peripheral blood mononuclear cells (PBMCs)