Learn More
Apexbio Technology LLC KT 5720 108068-98-0 50ug
SDP
Catalog No. 5000091324
Encompass_Preferred
KT 5720 (CAS 108068-98-0) is a cell-permeable reversible inhibitor of protein kinase A (PKA) that functions through ATP-competitive antagonism with an experimentally determined IC50 of 3 3 M This compound has been shown to counteract multidrug resistance in tumor cells mediated by MDR1 Additionally KT 5720 suppresses hyperpolarization-activated cyclic nucleotide-gated (HCN) cation channel activity resulting in decreased intracellular calcium levels and reduced excitability in dorsal root ganglion (DRG) neurons KT 5720 is utilized in studies investigating mechanisms underlying hematological malignancies and HCN or DRG-related disorders
Catalog No. 50-000-91324 Supplier Apexbio Technology LLC Supplier No. B90020.05
KT 5720 (CAS 108068-98-0) is a cell-permeable reversible inhibitor of protein kinase A (PKA) that functions through ATP-competitive antagonism with an experimentally determined IC50 of 3 3 M This compound has been shown to counteract multidrug resistance in tumor cells mediated by MDR1 Additionally KT 5720 suppresses hyperpolarization-activated cyclic nucleotide-gated (HCN) cation channel activity resulting in decreased intracellular calcium levels and reduced excitability in dorsal root ganglion (DRG) neurons KT 5720 is utilized in studies investigating mechanisms underlying hematological malignancies and HCN or DRG-related disorders