Learn More
Cayman Chemical KT 5720 Inhibitors
SDP
Supplier:  Cayman Chemical 10011011100 UG
Encompass_Preferred
A potent inhibitor of PKA; blocks PKA signaling through competitive inhibition of ATP (Ki = 60 nM); inhibits phosphorylase kinase, PDK1, MEK, MSK1, PKBα, and GSK3β
Catalog No. 50-288-8472