Learn More
StatLab KT Koverglass
SDP
Catalog No. 22130524
Change view
Click to view available options
Length (Metric):
50 mm
Catalog No. Length (Metric)
22-130-524 50 mm
1 options
This item is not returnable. View return policy
Catalog No. 22-130-524 Supplier StatLab Supplier No. KT102450
Add to Cart
Add to Cart
This item is not returnable. View return policy
KT Premium German Koverglass has a superior optical quality that ensures image clarity for microscopy and digital pathology
  • The superior raw materials provide Class 1 Chemical resistance, maintaining slides integrity for 20 plus years of archiving
  • Manufactured from SCHOTT D 263 M Glass
  • The state of the art machining process ensures that the edges are cut with high precision, which makes this coverglass especially suitable for automatic coverslipping machines
  • Specially designed glass 'dust' separates the cover glasses, preventing them from sticking together
  • These cover glasses are packed in plastic box with a hinged lid
  • Each of these plastic boxes contains 2 ounces, with 10 boxes per case for a total of 20 ounces per case
  • 24 x 50mm, No. 1 Thickness
Specifications
Product Type Coverslip
Shape Rectangle
Material Borosilicate Glass D 263 M
Length (Metric) 50 mm
Width (Metric) 24 mm