Learn More
Med Vet International Jorvet Equine Express Semen Transport Case, J0125
Catalog No. 5000495971
Encompass_Preferred
Transporting cooled stallion semen across long distances has been made possible by special transport shippers. This shipper has a cardboard outer shell with 2" thick Styrofoam liner. A special coolant pack is included along with two non-spermicidal 50cc syringes. The package has been approved by both FedEx and UPS. It also is a USDA-APHIS approved container for biological material. 14" x 8" x 10". 31/2 lbs.
Catalog No. 50-004-95971 Supplier Med Vet International Supplier No. JOR125
Transporting cooled stallion semen across long distances has been made possible by special transport shippers. This shipper has a cardboard outer shell with 2" thick Styrofoam liner. A special coolant pack is included along with two non-spermicidal 50cc syringes. The package has been approved by both FedEx and UPS. It also is a USDA-APHIS approved container for biological material. 14" x 8" x 10". 31/2 lbs.