Learn More
Med Vet International Stimuplex HNS 12 Complete Starter Kit, J1531, Each
Catalog No. 5000495654
Encompass_Preferred
The use of regional peripheral anesthesia is greatly increasing. Peripheral nerve stimulations (PNS) is used to guide the needle close to the nerve to facilitate a nerve blockage. The nerve is stimulated by low current with a motor response then a local anesthetic can be injected to effect. The special needles have dual channel tubing. One for electrical stimulation and the other for drug administration. The Braun Stimuplex is a small hand held device. The enlarged full graphic liquid crystal (LC) display gives you the important information at a glance. Kit includes 2 of 2 most popular needle sizes: J1531A Stimuplex Ultra 360 Needle, 22g x 3 1/4" (80mm) x 2 J1531B Stimuplex Ultra 360 Needle, 20g x 4" (100mm)
Catalog No. 50-004-95654 Supplier Med Vet International Supplier No. JOR1531
The use of regional peripheral anesthesia is greatly increasing. Peripheral nerve stimulations (PNS) is used to guide the needle close to the nerve to facilitate a nerve blockage. The nerve is stimulated by low current with a motor response then a local anesthetic can be injected to effect. The special needles have dual channel tubing. One for electrical stimulation and the other for drug administration. The Braun Stimuplex is a small hand held device. The enlarged full graphic liquid crystal (LC) display gives you the important information at a glance. Kit includes 2 of 2 most popular needle sizes: J1531A Stimuplex Ultra 360 Needle, 22g x 3 1/4" (80mm) x 2 J1531B Stimuplex Ultra 360 Needle, 20g x 4" (100mm)