Learn More
Med Vet International Steri Container, 8.25 x 2 x 2in, J0943X, Each
Catalog No. 5000495249
Encompass_Preferred
A small sterilizable kit that holds a few special instruments. This hinged-lid container fits into ultrasonic cleaners, plus is autoclavable.
Catalog No. 50-004-95249 Supplier Med Vet International Supplier No. JOR943X
A small sterilizable kit that holds a few special instruments. This hinged-lid container fits into ultrasonic cleaners, plus is autoclavable.