Learn More
Medchemexpress LLC HY-120177 5mg Medchemexpress, KT182 CAS:1402612-62-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1201775MG
Encompass_Preferred
Medchemexpress, HY-120177 5mg KT182 CAS:1402612-62-7 KT182 is a potent and selective inhibitor of α/β-hydrolase domain containing 6 (ABHD6), with an IC50 of 0.24 nM in Neuro2A cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9588