Learn More
Medchemexpress LLC HY-114926 10mg Medchemexpress, KT185 CAS:1472640-86-0 Purity:>98%
SDP
Catalog No. 501969376
Encompass_Preferred
Medchemexpress, HY-114926 10mg KT185 CAS:1472640-86-0 KT185 is an orally-bioavailable, brain-penetrant and selective ABHD6 inhibitor, with an IC50 0.21 nM in Neuro2A cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9376 Supplier Medchemexpress LLC Supplier No. HY11492610MG
Medchemexpress, HY-114926 10mg KT185 CAS:1472640-86-0 KT185 is an orally-bioavailable, brain-penetrant and selective ABHD6 inhibitor, with an IC50 0.21 nM in Neuro2A cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.