Learn More
Enzo Life Sciences KT5720 (100µg). CAS: 108068-98-0
SDP
Supplier:  Enzo Life Sciences BMLEI1990100
Encompass_Preferred
KT5720 is a hexylester derivative of K-252a and is a selective inhibitor of PKA (Ki=0.056 µM). Cell permeable. Purity: ≥98% (HPLC). Solubility: Soluble in methanol: clear colorless solution at 5mg/ml. Long Term Storage: -20°C.
Catalog No. 50-201-0608