Learn More
Medchemexpress LLC HY-N6789 50ug Medchemexpress, KT5720 CAS:108068-98-0 Purity:>98%
SDP
Catalog No. 502082963
Encompass_Preferred
Medchemexpress, HY-N6789 50ug KT5720 CAS:108068-98-0 KT5720 is a cell-permeable, potent, specific, reversible, ATP-competitive inhibitor of protein kinase A (PKA), with a Ki of 60 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2963 Supplier Medchemexpress LLC Supplier No. HYN678950UG
Medchemexpress, HY-N6789 50ug KT5720 CAS:108068-98-0 KT5720 is a cell-permeable, potent, specific, reversible, ATP-competitive inhibitor of protein kinase A (PKA), with a Ki of 60 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.