Learn More
Abcam KT5823, inhibitor of PKG, 100UG
Catalog No. 502707598
Encompass_Preferred
MW 495.5 Da, Purity >95%. Potent, selective, reversible inhibitor of Protein Kinase G (PKG) (Ki = 234 nM). Derivative of K252, selective over PKA and PKC (Ki values are >10 and 4 μM, respectively). Prevents increased phosphorylation of regulators of G-protein-signalling in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7598 Supplier Abcam Supplier No. AB120423100UG
MW 495.5 Da, Purity >95%. Potent, selective, reversible inhibitor of Protein Kinase G (PKG) (Ki = 234 nM). Derivative of K252, selective over PKA and PKC (Ki values are >10 and 4 μM, respectively). Prevents increased phosphorylation of regulators of G-protein-signalling in vivo.
The product is subject to the following: Abcam Restricted Use Statement