Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab KT5i+ Premium German Advanced Adhesion InkJet Compatible Slides
SDP

Catalog No. 21010333
Change view
Click to view available options
Color:
Aqua
Lavender
Orange
Edges:
90°, Clipped Corner
90°, Polished
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Edges
21-010-333 Orange 90°, Polished
21-010-329 Aqua 90°, Polished
21-010-330 Aqua 90°, Clipped Corner
21-010-331 Lavender 90°, Polished
21-010-332 Lavender 90°, Clipped Corner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-010-333 Supplier StatLab Supplier No. KT5PLUSIOR90
Add to Cart
Add to Cart

KT5i+ Premium German Advanced Adhesion InkJet Compatible Slides are compatible with both InkJet and UV laser printers and provide superb adhesion properties for holding the specimen on the slide permanently.

  • An excellent choice for H and E, special stains, frozen sections and even the toughest IHC procedures with difficult tissues
  • These slides feature superior adhesion, maintaining tissue integrity throughout the toughest of tissues or protocols
  • Tissue sections easily jump on the slide, indicating a hydrophobic waterbath behavior, but the precision manufacturing process ensures a 100% wettable surface for even reagent spreading
  • KT5+i stained slides will not have background staining
  • KT5+i slides provide a colored marking area for efficient specimen color-coding, identification, labeling and printing
  • KT5+i slides are available with 45° or 90° corners
  • KT5+i Slides are compatible with both inkjet and UV laser printers
  • Manufactured from premium German glass that enables superior clarity for digital pathology and imaging systems
  • Dimensions: 75.6 x 25.6 x 1 mm
  • 10 gross per case

Specifications

Length (Metric) 75.6 mm
Width (Metric) 25.6 mm
Type Adhesion Slide
Material High Quality Glass
Precleaned Yes
Thickness (Metric) 1 mm
Edges 90°, Polished
Frosted 1 End, 1 Side
Color Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.