Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KTC1101 | 2764833-47-6 | 98.1% | 476.48 | 100 MG
SDP

Catalog No. 5000392293
Encompass_Preferred

KTC1101 is an orally active pan-PI3K inhibitor that inhibits the PI3K signaling pathway, reducing downstream AKT and mTOR phosphorylation, and decreases Ki67 expression. Its anti-tumor effect stems from a dual mechanism: directly inhibiting tumor cell growth and dynamically enhancing immune response.

  • Induces G1 phase cell cycle arrest in various cell lines (0.1-50000 nM, 48 h).
  • Shows anti-proliferative activity (IC50 20-130 nM) without significant apoptosis.
  • Inhibits all PI3K isoforms (IC50s: PI3Kα 3.72 nM, PI3Kβ 36.29 nM, PI3Kδ 1.22 nM, PI3Kγ 17.09 nM).
  • Reduces AKT and mTOR phosphorylation in WB experiments (0-125 nM, 48 h).
  • Arrests tumor growth in mice xenografts (0-125 mg/kg/day for 14 days, p.o.) without recurrence.

Catalog No. 50-003-92293 Supplier Medchemexpress LLC Supplier No. HY162382100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KTC1101 is an orally active pan-PI3K inhibitor that inhibits the PI3K signaling pathway, reducing downstream AKT and mTOR phosphorylation, and decreases Ki67 expression. Its anti-tumor effect stems from a dual mechanism: directly inhibiting tumor cell growth and dynamically enhancing immune response.

  • Induces G1 phase cell cycle arrest in various cell lines (0.1-50000 nM, 48 h).
  • Shows anti-proliferative activity (IC50 20-130 nM) without significant apoptosis.
  • Inhibits all PI3K isoforms (IC50s: PI3Kα 3.72 nM, PI3Kβ 36.29 nM, PI3Kδ 1.22 nM, PI3Kγ 17.09 nM).
  • Reduces AKT and mTOR phosphorylation in WB experiments (0-125 nM, 48 h).
  • Arrests tumor growth in mice xenografts (0-125 mg/kg/day for 14 days, p.o.) without recurrence.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.