Learn More
Hach Company 2100Q Portable Turbidimeter Kit with USB and Power Module
Catalog No. 501997393 Shop All Hach Company Products
Encompass
The Hach 2100Q Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement. Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time.The USB+Power Module provides additional benefit by allowing the user to transfer data to a PC in XML format, transfer data to the Citizen PD-24 printer (2960100), operate the instrument on line power (100-240Vac, 50/60 HZ), charge NiMH batteries (2971304),perform firmware updates, and quickly enter Sample and User IDs with the optional barcode scanner (LZV566).
Catalog No. 50-199-7393 Supplier Hach Company Supplier No. 2100Q01USB
The Hach 2100Q Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement. Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time.The USB+Power Module provides additional benefit by allowing the user to transfer data to a PC in XML format, transfer data to the Citizen PD-24 printer (2960100), operate the instrument on line power (100-240Vac, 50/60 HZ), charge NiMH batteries (2971304),perform firmware updates, and quickly enter Sample and User IDs with the optional barcode scanner (LZV566).