Learn More
Hach Company 2100Q IS with USB+Power Module
Catalog No. 501997395 Shop All Hach Company Products
Encompass
The Hach 2100Q IS Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement. Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time.The USB+Power Module provides additional benefit by allowing the user to transfer data to a PC in XML format, transfer data to the Citizen PD-24 printer (2960100), operate the instrument on line power (100-240Vac, 50/60 HZ), charge NiMH batteries (2971304),perform firmware updates, and quickly enter Sample and User IDs with the optional barcode scanner (LZV566).
Catalog No. 50-199-7395 Supplier Hach Company Supplier No. 2100QIS01USB
The Hach 2100Q IS Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement. Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time.The USB+Power Module provides additional benefit by allowing the user to transfer data to a PC in XML format, transfer data to the Citizen PD-24 printer (2960100), operate the instrument on line power (100-240Vac, 50/60 HZ), charge NiMH batteries (2971304),perform firmware updates, and quickly enter Sample and User IDs with the optional barcode scanner (LZV566).