Learn More
Hach Company Acid Reagent Powder Pillows, 25 mL, pk/1000
Supplier:  Hach Company 104228
Encompass
For determination of high range silica by the silicomolybdate method (Hach method 8185) Range: 1-100.0 mg/L SiO2. Also used in the iodometric method (Hach method 10100) for the determination of high range hypchlorite (50-150 g/L as Cl2). Pack of 1000 powder pillows.
Catalog No. 50-199-4497