Learn More
Hach Company Amino Acid F Reagent Powder Pillows, 25 mL, pk/1000
Supplier:  Hach Company 2253828
Encompass
Reagent for Silica determination by the Heteropoly Blue method. Hach Method 8186. Sample Size: 25 mL. Pack of 1000 Powder Pillows
Catalog No. 50-199-5639