Learn More
Hach Company Amino Acid F Reagent for S5000 Silica (2.7 L)
Supplier:  Hach Company 2353103
Encompass
Amino Acid F Reagent (2.7 L) used in the Series 5000 Silica Analyzer. Amino Acid F reagent is supplied in two parts to ensure a long shelf life: a 3-liter bottle of Amino Acid F Solution and a bottle of Amino Acid F Powder. Powder must be added to the solution immediately before installation in the analyzer.
Catalog No. 50-199-5868