Learn More
Hach Company BART Test for Iron-Related Bacteria, pk/27
Supplier:  Hach Company 2432327
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence of process disruptors, such as iron-reducing bacteria (IRB) that could be eating your pipes. An important agent involved in biofouling, IRB can cause corrosion, clogging, fouling, and hygiene risk.
Catalog No. 50-199-5976