Learn More
Hach Company BART Test for Sulfate-Reducing Bacteria, pk/27
Supplier:  Hach Company 2432427
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Sulfate Reducing Bacteria (SRB) are one of the most important agents in biofouling and can cause corrosion, clogging, fouling of the water, and increased hygiene risks.
Catalog No. 50-199-5978