Learn More
Hach Company HQ440D Water Quality Laboratory Biochemical Oxygen Demand (BOD) Meter Package with Optical LBOD101 Sensor
Catalog No. 501997263 Shop All Hach Company Products
Encompass
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD meters feature a simple user interface that does not require manuals or training to operate. The Intellical LBOD101 is the latest embodiment of Hach’s patented Luminescent Dissolved Oxygen (LDO) technology. The LBOD101 is a digital, luminescent dissolved oxygen (LDO) sensor with a replaceable, integrated stirring system. The LBOD101 measures the dissolved oxygen for BOD (Biochemical Oxygen Demand) determination in BOD bottles in BOD5 testing applications. The LBOD101 electrode is used for initial and final dissolved oxygen measurements.
Catalog No. 50-199-7263 Supplier Hach Company Supplier No. 8508500
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD meters feature a simple user interface that does not require manuals or training to operate. The Intellical LBOD101 is the latest embodiment of Hach’s patented Luminescent Dissolved Oxygen (LDO) technology. The LBOD101 is a digital, luminescent dissolved oxygen (LDO) sensor with a replaceable, integrated stirring system. The LBOD101 measures the dissolved oxygen for BOD (Biochemical Oxygen Demand) determination in BOD bottles in BOD5 testing applications. The LBOD101 electrode is used for initial and final dissolved oxygen measurements.