Learn More
Hach Company Ammonia, Free & Total, and Monochloramine Chemkey Reagents (box of 25 each)
Catalog No. 501997278 Shop All Hach Company Products
Encompass
Get two measurements (Free Ammonia and Total Ammonia) on one Chemkey. By measuring two parameters on one Chemkey free up a slot and save time. Must use the Free & Total Ammonia Chemkey along with the Monochloramine Chemkey.Ammonia, free 0.05 - 0.5 mg/L NH3-N / total 0.05 - 1.50 mg/L NH3-N, and Monochloramine, 0.04 - 4.0 mg/L Cl2, Chemkeys. For SL1000 Portable Parallel Analyzer (PPA).
Catalog No. 50-199-7278 Supplier Hach Company Supplier No. 8791900
Get two measurements (Free Ammonia and Total Ammonia) on one Chemkey. By measuring two parameters on one Chemkey free up a slot and save time. Must use the Free & Total Ammonia Chemkey along with the Monochloramine Chemkey.Ammonia, free 0.05 - 0.5 mg/L NH3-N / total 0.05 - 1.50 mg/L NH3-N, and Monochloramine, 0.04 - 4.0 mg/L Cl2, Chemkeys. For SL1000 Portable Parallel Analyzer (PPA).