Learn More
Hach Company Total Chlorine Chemkey Reagents (Qty 300)
Supplier:  Hach Company 8499400
Encompass
Total Chlorine Chemkeys, range 0.04 - 10 mg/L Cl2. For SL1000 Portable Parallel Analyzer (PPA). 12 boxes of 25 (300 tests total quantity).USEPA accepted for reporting in drinking water analysis using Hach Method 10260.
Catalog No. 50-199-7226