Learn More
Hach Company ChromaVer 3 Chromium Reagent Powder Pillows, 5 or 10 mL, pk/1000
Catalog No. 501994878 Shop All Hach Company Products
Encompass
For determination of hexavalent chromium by the Diphenylcarbohydrazide method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L. Kit Sample Size: 5 mL. Kit Range: Up to 1.50 Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4878 Supplier Hach Company Supplier No. 1271028
For determination of hexavalent chromium by the Diphenylcarbohydrazide method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L. Kit Sample Size: 5 mL. Kit Range: Up to 1.50 Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.