Learn More
Hach Company Citric Acid Reagent Powder Pillows for Silica, 10 mL, pk/1000
Catalog No. 501995407 Shop All Hach Company Products
Encompass
For determination of high range silica by the silicomolybdate method (Hach method 8185), and low range silica by the heteropoly blue method (Hach method 8186). Pack of 1000 Powder Pillows.
Catalog No. 50-199-5407 Supplier Hach Company Supplier No. 2106228
For determination of high range silica by the silicomolybdate method (Hach method 8185), and low range silica by the heteropoly blue method (Hach method 8186). Pack of 1000 Powder Pillows.